Learn More
Enterprise Technology Solutions Light Duty Wiping Paper Towel, 4.5" X 8.25", White, 16,800/Box
SDP
Supplier:  Enterprise Technology Solutions NSN7218884
Encompass_Preferred
ABILITYONE 7920007218884 Skilcraft Cleans delicate instruments and lenses in medical research, electronics and lab areas. Great for scientific cleaning applications requiring low lint, soft tissue for take wiping and cleaning with low abrasion. Cleans glass without streaking. Handy pop-up dispensers save space.
Catalog No. 50-204-1147