Learn More
Enterprise Technology Solutions Scrubwipes Preparation Wipers, Blue, 300/Carton
SDP
Catalog No. 502040101
Encompass_Preferred
ABILITYONE 7920012330483 Skilcraft Low-lint wipers designed for use with chemicals. Can be used with solvents, inks and paints. Ideal choice for industrial applications using lubricants, oil and grease.
Catalog No. 50-204-0101 Supplier Enterprise Technology Solutions Supplier No. NSN2330483
ABILITYONE 7920012330483 Skilcraft Low-lint wipers designed for use with chemicals. Can be used with solvents, inks and paints. Ideal choice for industrial applications using lubricants, oil and grease.