Learn More
Cayman Chemical 7a 25dIhydroxy Cholesterol 5mg
SDP
Catalog No. 502951387
Encompass_Preferred
A potent agonist of GPR183 that induces specific activation of the receptor with an EC50 value of 50 nM (Kd = 450 pM); acts as a potent chemokine, affecting migration of immune cells expressing GPR183 both in vitro (EC50 = 500 pM) and in vivo
Catalog No. 50-295-1387 Supplier Cayman Chemical Supplier No. 110325 MG
Add to Cart
Add to Cart
A potent agonist of GPR183 that induces specific activation of the receptor with an EC50 value of 50 nM (Kd = 450 pM); acts as a potent chemokine, affecting migration of immune cells expressing GPR183 both in vitro (EC50 = 500 pM) and in vivo