Learn More
Cayman Chemical 7α-hydroxy Cholesterol, Purity: ≥98%, CAS: 566-26-7, 10 mg
SDP
Catalog No. 502981603
Encompass_Preferred
An oxysterol and a precursor in the biosynthesis of CA and CDCA; formed via the oxidation of cholesterol by CYP7A1 in rat liver microsomes; increases ICAM-1, VCAM-1, and E-selectin levels in HUVECs at 40 µM; increases secretion of CCL2 and MMP-9 in serum-deprived THP-1 cells at 5 µg/ml; has been found in macrophages isolated from atherosclerotic lesions in rabbits fed a high-cholesterol diet
Catalog No. 50-298-1603 Supplier Cayman Chemical Supplier No. 2009810 MG
An oxysterol and a precursor in the biosynthesis of CA and CDCA; formed via the oxidation of cholesterol by CYP7A1 in rat liver microsomes; increases ICAM-1, VCAM-1, and E-selectin levels in HUVECs at 40 µM; increases secretion of CCL2 and MMP-9 in serum-deprived THP-1 cells at 5 µg/ml; has been found in macrophages isolated from atherosclerotic lesions in rabbits fed a high-cholesterol diet