Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids 7α-Hydroxycholesterol, Powder, 10 mg

Catalog No. NC2827738
Encompass

7α-Hydroxycholesterol (cholest-5-en-3β,7α-diol) is a highly pure lipid used in scientific research to explore its various biological roles, including its function in bile acid synthesis and its contribution to immune responses. This compound is provided as a stable powder and is suitable for a range of applications in the study of lipid standards.

  • High purity lipid with over 99% purity
  • Physical state: Powder
  • Molecular formula: C27H46O2
  • Storage temperature: -20°C
  • Stability: 1 year
  • Non-hygroscopic and non-light sensitive
  • Packaged as 1 x 10 mg
  • Shipped in dry ice

Catalog No. NC2827738 Supplier Avanti Polar Lipids Supplier No. 700034P10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7α-Hydroxycholesterol (cholest-5-en-3β,7α-diol) is a highly pure lipid used in scientific research to explore its various biological roles, including its function in bile acid synthesis and its contribution to immune responses. This compound is provided as a stable powder and is suitable for a range of applications in the study of lipid standards.

  • High purity lipid with over 99% purity
  • Physical state: Powder
  • Molecular formula: C27H46O2
  • Storage temperature: -20°C
  • Stability: 1 year
  • Non-hygroscopic and non-light sensitive
  • Packaged as 1 x 10 mg
  • Shipped in dry ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.