Learn More
Cayman Chemical 7AADCFSE CelMedIATd Cyto 1ea
SDP
Catalog No. 502896436
Encompass_Preferred
Cayman’s 7-AAD/CFSE Cell-Mediated Cytotoxicity Assay Kit employs CFSE to label target cells within the mixed cell population and 7-AAD to label dead cells. This kit provides an improvement over the traditional 51chromium (51Cr) release assay to assess cell-mediated cytotoxicity. CFSE labeling is more sensitive, does not employ radioisotopes, and cytolysis can be assessed at the single-cell level. The mixed lymphocyte reaction is a cornerstone method of immunology{17323}, and this kit can help immunologists to detect subtle changes in cytotoxic lymphocyte function. The kit provides sufficient reagents to effectively stain approximately 109 cells.
Catalog No. 50-289-6436 Supplier Cayman Chemical Supplier No. 6001201 EA
Cayman’s 7-AAD/CFSE Cell-Mediated Cytotoxicity Assay Kit employs CFSE to label target cells within the mixed cell population and 7-AAD to label dead cells. This kit provides an improvement over the traditional 51chromium (51Cr) release assay to assess cell-mediated cytotoxicity. CFSE labeling is more sensitive, does not employ radioisotopes, and cytolysis can be assessed at the single-cell level. The mixed lymphocyte reaction is a cornerstone method of immunology{17323}, and this kit can help immunologists to detect subtle changes in cytotoxic lymphocyte function. The kit provides sufficient reagents to effectively stain approximately 109 cells.