Learn More
Selleck Chemical LLC 7ACC2
SDP
Catalog No. 5000002905
Encompass_Preferred
7ACC2 is a potent inhibitor of mitochondrial pyruvate transport 1 (MCT1) with an IC50 value of 10 nM for lactate uptake in SiHa human cervix carcinoma cells
Catalog No. 50-000-02905 Supplier Selleck Chemical LLC Supplier No. S67322MG
7ACC2 is a potent inhibitor of mitochondrial pyruvate transport 1 (MCT1) with an IC50 value of 10 nM for lactate uptake in SiHa human cervix carcinoma cells