Learn More
Cayman Chemical 7ahydroxy DehydroepIndROS 10mg
SDP
Catalog No. 502948474
Encompass_Preferred
An active metabolite of dehydroepiandrosterone; formed from dehydroepiandrosterone via 7-hydroxylation by CYPs; increases antigen-induced production of anti-tetanus toxoid and anti-pertussis antigen IgGs in co-cultures of human tonsil-derived B and T cells at 1 µM; increases serum concentrations of anti-lysozyme IgG in mice at 6.25 mg/kg
Catalog No. 50-294-8474 Supplier Cayman Chemical Supplier No. 3426810 MG
An active metabolite of dehydroepiandrosterone; formed from dehydroepiandrosterone via 7-hydroxylation by CYPs; increases antigen-induced production of anti-tetanus toxoid and anti-pertussis antigen IgGs in co-cultures of human tonsil-derived B and T cells at 1 µM; increases serum concentrations of anti-lysozyme IgG in mice at 6.25 mg/kg