Learn More
Cayman Chemical 7α-hydroxy-3-oxo-5β-Cholan-24-oic Acid, Purity: ≥95%, CAS: 4185-00-6, 10 mg
SDP
Catalog No. 502916323
Encompass_Preferred
A metabolite of chenodeoxycholic acid; formed from chenodeoxycholic acid primarily by CYP3A4 but also by CYP3A5 and other P450 enzymes at lower rates
Catalog No. 50-291-6323 Supplier Cayman Chemical Supplier No. 3815910 MG
A metabolite of chenodeoxycholic acid; formed from chenodeoxycholic acid primarily by CYP3A4 but also by CYP3A5 and other P450 enzymes at lower rates