Learn More
TARGETMOL CHEMICALS INC 7alpha-Hydroxy-4-cholesten
SDP
Catalog No. 502880652
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 7alpha-Hydroxy-4-cholesten-3-one is an intermediate in bile acid synthesis from cholesterol, a PXR agonist, and a biomarker for bile acid loss and diseases related to defective bile acid biosynthesis. It is also the physiological substrate for [CYP8B1]. Purity 99.14%
Catalog No. 50-288-0652 Supplier TARGETMOL CHEMICALS INC Supplier No. T101961MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 7alpha-Hydroxy-4-cholesten-3-one is an intermediate in bile acid synthesis from cholesterol, a PXR agonist, and a biomarker for bile acid loss and diseases related to defective bile acid biosynthesis. It is also the physiological substrate for [CYP8B1]. Purity 99.14%