Learn More
AdipoGen 7b-OH-cholesteryl-bishemisucci
SDP
Catalog No. 502560712
Encompass_Preferred
Chemical. CAS 105449-93-2. Formula C35H54O8 . 2C2H7NO. MW 602.80 . 61.08. Synthetic Water-soluble derivatives of 7beta-hydroxycholesterol. Anticancer compound, inducing apoptosis in various solid tumor cell lines.
Catalog No. 50-256-0712 Supplier AdipoGen Supplier No. CDXH0123M050
Chemical. CAS 105449-93-2. Formula C35H54O8 . 2C2H7NO. MW 602.80 . 61.08. Synthetic Water-soluble derivatives of 7beta-hydroxycholesterol. Anticancer compound, inducing apoptosis in various solid tumor cell lines.