Learn More
Cayman Chemical 7Benzyloxy4 trIfluoromthy 10mg
SDP
Catalog No. 502910303
Encompass_Preferred
A fluorogenic substrate for CYP1A2 and CYP3A; HFC is released upon enzymatic cleavage by CYP1A2 and CYP3A and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-291-0303 Supplier Cayman Chemical Supplier No. 3572110 MG
A fluorogenic substrate for CYP1A2 and CYP3A; HFC is released upon enzymatic cleavage by CYP1A2 and CYP3A and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively