Learn More
Cayman Chemical 7Benzyloxy4 trIfluoromthy 50mg
SDP
Catalog No. 502910403
Encompass_Preferred
A fluorogenic substrate for CYP1A2 and CYP3A; HFC is released upon enzymatic cleavage by CYP1A2 and CYP3A and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-291-0403 Supplier Cayman Chemical Supplier No. 3572150 MG
A fluorogenic substrate for CYP1A2 and CYP3A; HFC is released upon enzymatic cleavage by CYP1A2 and CYP3A and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively