Learn More
AdipoGen 7beta-Hydroxycholesterol
SDP
Supplier:  AdipoGen CDXH0116M100
Encompass_Preferred
Chemical. CAS 566-27-8. Formula C27H46O2. MW 402.65. Synthetic Oxysterole. Cholesterol oxidation metabolite. Induces cell death/apoptosis and shows antiproliferative activity against cancer cells, involving mitochondrial perturbation, oxidative stress and lysosomal destabilization. Its membrane organizing properties may have implications in Alzheimer’s disease. Increased levels correlate with risk of cardiovascular diseases including atherosclerosis. Shown to have proinflammatory activity.
Catalog No. 50-256-0718