Learn More
AdipoGen 7BIO
SDP
Supplier:  AdipoGen AGMRC0020M010
Encompass_Preferred
Chemical. CAS 916440-85-2. Formula C16H10BrN3O2. MW 356.2. Anticancer compound. Caspase-independent, non-apoptotic cell death inducer. ATP-competitive Flt3 inhibitor. Necrosis inducer. Aurora B and C kinase inhibitor. DYRK1A and DYRK 2 Dual-specificity tyrosine phosphorylation-regulated kinase inhibitor. Inflammasome activator.
Catalog No. 50-256-2162