Learn More
AdipoGen 7BIO (50 mg)
SDP
Supplier:  AdipoGen AGMRC0020M050
Encompass_Preferred
Chemical. CAS: 916440-85-2. Formula: C16H10BrN3O2. MW: 356.2. Anticancer compound. Caspase-independent, non-apoptotic cell death inducer. ATP-competitive Flt3 inhibitor. Necrosis inducer. Aurora B and C kinase inhibitor. DYRK1A and DYRK 2 (Dual-specificity tyrosine phosphorylation-regulated kinase) inhibitor. Inflammasome activator.
Catalog No. 50-205-4240