Learn More
Cayman Chemical 7-Deaza-7-iodo-2'-deoxyguanosine, Purity: ≥98%, CAS: 172163-62-1, 500 mg
SDP
Catalog No. 502944097
Encompass_Preferred
A nucleoside derivative; has been used as an intermediate in the synthesis of fluorescent nucleotide derivatives for incorporation into single- or double-stranded oligonucleotides
Catalog No. 50-294-4097 Supplier Cayman Chemical Supplier No. 40643500 MG
A nucleoside derivative; has been used as an intermediate in the synthesis of fluorescent nucleotide derivatives for incorporation into single- or double-stranded oligonucleotides