Learn More
Cayman Chemical 7deaza8bromo NAD sodIM 700ug
SDP
Catalog No. 502916800
Encompass_Preferred
A derivative of NAD+ and a synthetic intermediate; has been used as an intermediate in the synthesis of cyclic and membrane-permeable diphosphate modulators of TRPM2
Catalog No. 50-291-6800 Supplier Cayman Chemical Supplier No. 38479700 UG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A derivative of NAD+ and a synthetic intermediate; has been used as an intermediate in the synthesis of cyclic and membrane-permeable diphosphate modulators of TRPM2