Learn More
Cayman Chemical 7DeazaadenosIn5OtrIphos 2.5mg
SDP
Catalog No. 502918069
Encompass_Preferred
A modified nucleotide and an active metabolite of tubercidin; incorporated into the DNA of mouse L-cells using DNA polymerase α but not into the RNA of E. coli using RdRps; has been used as a probe for the ATP binding site of PKA
Catalog No. 50-291-8069 Supplier Cayman Chemical Supplier No. 383772.5 MG
A modified nucleotide and an active metabolite of tubercidin; incorporated into the DNA of mouse L-cells using DNA polymerase α but not into the RNA of E. coli using RdRps; has been used as a probe for the ATP binding site of PKA