Learn More
Cayman Chemical 7-Diethylamino-3-(4-maleimidophenyl)-4-methylcoumarin, Purity: ≥95%, CAS: 76877-33-3, 10 mg
SDP
Catalog No. 502965173
Encompass_Preferred
A thiol-reactive fluorescent probe; fluorescence intensity increases when bound to cysteine residues ; ex/em = 387/468, respectively
Catalog No. 50-296-5173 Supplier Cayman Chemical Supplier No. 1820910 MG
A thiol-reactive fluorescent probe; fluorescence intensity increases when bound to cysteine residues ; ex/em = 387/468, respectively