Learn More
Cayman Chemical 7Ethoxy4 trIfluoromethylc 10mg
SDP
Catalog No. 502948844
Encompass_Preferred
A fluorogenic substrate for CYPs; HFC is released upon O-deethylation by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-294-8844 Supplier Cayman Chemical Supplier No. 3461010 MG
A fluorogenic substrate for CYPs; HFC is released upon O-deethylation by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively