Learn More
Cayman Chemical 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonate (ammonium salt), Purity: ≥98%, CAS: 84806-27-9, 250 mg
SDP
Catalog No. 502947919
Encompass_Preferred
A thiol-reactive fluorogenic probe; has been used to quantify homocysteine, cysteine, and cysteamine levels in human plasma; ex/em = 380/515 nm, respectively
Catalog No. 50-294-7919 Supplier Cayman Chemical Supplier No. 34564250 MG
A thiol-reactive fluorogenic probe; has been used to quantify homocysteine, cysteine, and cysteamine levels in human plasma; ex/em = 380/515 nm, respectively