Learn More
Medchemexpress LLC 7h-Indeno[2,1-c]quinolin-7-one, 6-[[2-(dimethylamino)ethyl)amino]-3-hydroxy-, hydrochloride (1:2) | 174634-09-4 | 98.5% | 406.31 | 25 MG
SDP
Catalog No. 5000350558
Encompass_Preferred
TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II. It is used for cancer research and is intended for research use only.
  • Dual inhibitor of DNA topoisomerase I/II
  • Utilized in cancer research
  • Active on CCRF-CEM cells with an IC50 of 5 nM
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells
  • Inhibits viability of HeLa cells with an IC50 of 40 nM
  • Causes significant tumor growth suppression in mice bearing Lewis lung carcinoma (LLC) cells
Catalog No. 50-003-50558 Supplier Medchemexpress LLC Supplier No. 25MGHY13758A
TAS-103 dihydrochloride is a dual inhibitor of DNA topoisomerase I/II. It is used for cancer research and is intended for research use only.
  • Dual inhibitor of DNA topoisomerase I/II
  • Utilized in cancer research
  • Active on CCRF-CEM cells with an IC50 of 5 nM
  • Highly cytotoxic to Lewis lung carcinoma (LLC) cells
  • Inhibits viability of HeLa cells with an IC50 of 40 nM
  • Causes significant tumor growth suppression in mice bearing Lewis lung carcinoma (LLC) cells