Learn More
Medchemexpress LLC 7H-pyrazolo[4,3-d]pyrimidin-7-one, 3-[4-(aminomethyl)phenyl]-2-methyl-6-(piperidin-4-yl)methyl... | 2202738-42-7 | MFCD31813656 | 99.1% | 568.59 g·mol⁻¹ | C29H31F3N6O3 | 5 MG
SDP
Catalog No. 5000263603
Encompass_Preferred
USP7-IN-3 is a selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7) used for biochemical and cell-based research, including studies of p53/MDM2 signaling and acute lymphoblastic leukemia. It is supplied as a white to off-white solid with characterized physicochemical and biological properties for in vitro and in vivo use.
  • Selective allosteric inhibition of USP7.
  • Reduces MDM2 levels and increases p53 expression in cell assays.
  • Inhibits proliferation of RS4;11 acute lymphoblastic leukemia cells.
  • High purity (~99.1%) with defined molecular weight and formula.
  • Soluble in DMSO; in vivo formulation protocols documented.
  • Stable as powder at -20°C; solution storage conditions specified.
Catalog No. 50-002-63603 Supplier Medchemexpress LLC Supplier No. HY1121285MG
Add to Cart
Add to Cart
USP7-IN-3 is a selective allosteric inhibitor of ubiquitin-specific protease 7 (USP7) used for biochemical and cell-based research, including studies of p53/MDM2 signaling and acute lymphoblastic leukemia. It is supplied as a white to off-white solid with characterized physicochemical and biological properties for in vitro and in vivo use.
  • Selective allosteric inhibition of USP7.
  • Reduces MDM2 levels and increases p53 expression in cell assays.
  • Inhibits proliferation of RS4;11 acute lymphoblastic leukemia cells.
  • High purity (~99.1%) with defined molecular weight and formula.
  • Soluble in DMSO; in vivo formulation protocols documented.
  • Stable as powder at -20°C; solution storage conditions specified.