Learn More
Medchemexpress LLC 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-(2-ethoxyphenyl)-1,6-dihydro-1-methyl-3-propyl- | 139756-21-1 | 99.8% | 312.37 | 250 MG
SDP
Catalog No. 5000451606
Encompass_Preferred
PDE5-IN-7 (compound 8) is a selective phosphodiesterase 5 (PDE 5) inhibitor with an IC50 value of 5 nM, while having an IC50 of 300 nM for PDE 1. It can be detected and isolated from Sildenafil derivatives. It is intended for research use only and is not sold to patients.
  • Selective phosphodiesterase 5 (PDE 5) inhibitor
  • IC50 value of 5 nM for PDE5
  • IC50 value of 300 nM for PDE1
  • Detected and isolated from Sildenafil derivatives
Catalog No. 50-004-51606 Supplier Medchemexpress LLC Supplier No. 250MGHYW011336
PDE5-IN-7 (compound 8) is a selective phosphodiesterase 5 (PDE 5) inhibitor with an IC50 value of 5 nM, while having an IC50 of 300 nM for PDE 1. It can be detected and isolated from Sildenafil derivatives. It is intended for research use only and is not sold to patients.
  • Selective phosphodiesterase 5 (PDE 5) inhibitor
  • IC50 value of 5 nM for PDE5
  • IC50 value of 300 nM for PDE1
  • Detected and isolated from Sildenafil derivatives