Learn More
Medchemexpress LLC 7H-pyrrolo[2,3-d]pyrimidin-4-amine, N-butyl-N-ethyl-2,5,6-trimethyl-7-(trimethylphenyl) hydrochloride | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g/mol | C24H35ClN4 | 50 MG
SDP
Catalog No. 5000244550
Encompass_Preferred
Antalarmin hydrochloride is a research-grade, non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist supplied as a white to light yellow solid for preclinical research. It is used to study stress-response pathways, CRH/CRF signaling, anxiety-like behavior, and inflammation models. The compound is provided at high purity and supplied in small mg-scale pack sizes suitable for laboratory use.
  • Non-peptide CRHR1 antagonist used in stress and neuroendocrine research.
  • High purity for reliable experimental results.
  • Solid form, easy to weigh and formulate.
  • Available in mg-scale pack sizes for preclinical studies.
  • Includes datasheet, certificate of analysis, and safety data sheet documentation.
Catalog No. 50-002-44550 Supplier Medchemexpress LLC Supplier No. HY10337750MG
Antalarmin hydrochloride is a research-grade, non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist supplied as a white to light yellow solid for preclinical research. It is used to study stress-response pathways, CRH/CRF signaling, anxiety-like behavior, and inflammation models. The compound is provided at high purity and supplied in small mg-scale pack sizes suitable for laboratory use.
  • Non-peptide CRHR1 antagonist used in stress and neuroendocrine research.
  • High purity for reliable experimental results.
  • Solid form, easy to weigh and formulate.
  • Available in mg-scale pack sizes for preclinical studies.
  • Includes datasheet, certificate of analysis, and safety data sheet documentation.