Learn More
Cayman Chemical 7hydroxy CLOrpromazInhyd 2.5mg
SDP
Catalog No. 502947398
Encompass_Preferred
An active metabolite of chlorpromazine; increases plasma prolactin levels in male rats at 5 mg/kg; reverses D-amphetamine-induced depression of the dopaminergic neuronal firing rate in the rat substantia nigra in rats at 1.6 mg/kg
Catalog No. 50-294-7398 Supplier Cayman Chemical Supplier No. 337292.5 MG
An active metabolite of chlorpromazine; increases plasma prolactin levels in male rats at 5 mg/kg; reverses D-amphetamine-induced depression of the dopaminergic neuronal firing rate in the rat substantia nigra in rats at 1.6 mg/kg