Learn More
Cayman Chemical 7hydroxy CoumarIn sulfatep
SDP
Catalog No. 502980647
Encompass_Preferred
A phase II metabolite of coumarin; an internal standard for coumarin metabolism
Catalog No. 50-298-0647 Supplier Cayman Chemical Supplier No. 20892500 UG
Add to Cart
Add to Cart
A phase II metabolite of coumarin; an internal standard for coumarin metabolism