Learn More
Cayman Chemical 7-hydroxycoumarinyl Arachidonate, Purity: ≥98%, CAS: 161180-11-6, 10 mg
SDP
Catalog No. 502893544
Encompass_Preferred
A fluorogenic substrate for cPLA2; hydrolysis results in the release of the fluorescent compound 7-hydroxycoumarin (ex 335 nm, em 450 nm)
Catalog No. 50-289-3544 Supplier Cayman Chemical Supplier No. 6291010 MG
A fluorogenic substrate for cPLA2; hydrolysis results in the release of the fluorescent compound 7-hydroxycoumarin (ex 335 nm, em 450 nm)