Learn More
Cayman Chemical 7-hydroxycoumarinyl Arachidonate, Purity: ≥98%, CAS: 161180-11-6, 50 mg
SDP
Catalog No. 502893258
Encompass_Preferred
A fluorogenic substrate for cPLA2; hydrolysis results in the release of the fluorescent compound 7-hydroxycoumarin (ex 335 nm, em 450 nm)
Catalog No. 50-289-3258 Supplier Cayman Chemical Supplier No. 6291050 MG
A fluorogenic substrate for cPLA2; hydrolysis results in the release of the fluorescent compound 7-hydroxycoumarin (ex 335 nm, em 450 nm)