Learn More
Sklar Instruments Single Use, Econo-Sterile, Multi-Cut Utility Scissors, Sklar 7 in., Blunt/Blunt, Angled, Serrated/Smooth, Stainless Steel / Plastic, Disposable. CS of 50.
SDP
Catalog No. 501094354 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Multi-Cut Utility Scissors, Sklar, 7 in., Blunt/Blunt, Angled, Serrated/Smooth, Stainless Steel / Plastic, Disposable. CS of 50. Econo Sterile Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes.
Catalog No. 50-109-4354 Supplier Sklar Instruments Supplier No. 962704
Single Use, Econo-Sterile, Multi-Cut Utility Scissors, Sklar, 7 in., Blunt/Blunt, Angled, Serrated/Smooth, Stainless Steel / Plastic, Disposable. CS of 50. Econo Sterile Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes.