Learn More
Sklar Instruments Scissors, Merit Metzenbaum Dissecting Scissors, Sklar Blunt/Blunt, 7 in., Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094121 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Merit Metzenbaum Dissecting Scissors, Sklar, Blunt/Blunt, 7 in., Straight, Smooth, Stainless Steel, EA. Merit Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting fine tissue. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
Catalog No. 50-109-4121 Supplier Sklar Instruments Supplier No. 97339
Scissors, Merit Metzenbaum Dissecting Scissors, Sklar, Blunt/Blunt, 7 in., Straight, Smooth, Stainless Steel, EA. Merit Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting fine tissue. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.