Learn More
Cole-Parmer Nylon Micro Brush, 4" Handle, 3"L x 00.1875-0.5" Dia Brush; 12/Pk
Supplier:  Cole-Parmer 8455195
Encompass
Brush design is ideal for cleaning 2- to 100-ml centrifuge tubes, micro tubes, and a variety of other containers.
Catalog No. NC0028017