Learn More
Sklar Instruments Scissors, Operating Scissors, Sklar, 7 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Reusable, EA.
SDP
Catalog No. 501093376 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Operating Scissors, Sklar, 7 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Reusable, EA. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is curved with smooth, blunt/blunt tips and a length of 7 inches.
Catalog No. 50-109-3376 Supplier Sklar Instruments Supplier No. 152070
Scissors, Operating Scissors, Sklar, 7 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Reusable, EA. Sklar Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. This product is curved with smooth, blunt/blunt tips and a length of 7 inches.