Learn More
Sklar Instruments Scissors, Surgi-OR Metzenbaum Dissecting Scissors, Sklar 7 in., Blunt/Blunt,Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094071 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Surgi-OR Metzenbaum Dissecting Scissors, Sklar, 7 in., Blunt/Blunt,Curved, Smooth, Stainless Steel, EA. Surgi-OR Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product has straight, smooth, blunt tips, and an overall length of 7 inches.
Catalog No. 50-109-4071 Supplier Sklar Instruments Supplier No. 95340
Scissors, Surgi-OR Metzenbaum Dissecting Scissors, Sklar, 7 in., Blunt/Blunt,Curved, Smooth, Stainless Steel, EA. Surgi-OR Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product has straight, smooth, blunt tips, and an overall length of 7 inches.