Learn More
Cayman Chemical 7-keto Dehydroepiandrosterone, Purity: ≥98%, CAS: 566-19-8, 10 mg
SDP
Catalog No. 502964126
Encompass_Preferred
An analytical reference standard categorized as a steroid metabolite of dehydroepiandrosterone; reduces ethanol consumption without the hormone-generating effects of dehydroepiandrosterone in rats; intended for research and forensic applications
Catalog No. 50-296-4126 Supplier Cayman Chemical Supplier No. 2249710 MG
An analytical reference standard categorized as a steroid metabolite of dehydroepiandrosterone; reduces ethanol consumption without the hormone-generating effects of dehydroepiandrosterone in rats; intended for research and forensic applications