Learn More
Cayman Chemical 7keto25hydroxy CholesTRol 5mg
SDP
Catalog No. 502933483
Encompass_Preferred
An oxysterol; a proposed metabolite of 7-keto cholesterol and 7-dehydro cholesterol; binds to and activates Smo; increases EGF high affinity binding site numbers in NRK cells
Catalog No. 50-293-3483 Supplier Cayman Chemical Supplier No. 259735 MG
An oxysterol; a proposed metabolite of 7-keto cholesterol and 7-dehydro cholesterol; binds to and activates Smo; increases EGF high affinity binding site numbers in NRK cells