Learn More
Chemglass Life Sciences Chemglass Jacketed Glass Bioreactor Vessel, 7 L (CLS-1384-01)
SDP
Catalog No. NC2157841
Encompass
The Chemglass Jacketed Glass Bioreactor Vessel is a 7-liter jacketed borosilicate glass vessel designed for laboratory bioprocessing and fermentation. It provides jacketed temperature control, clear graduated sightlines, and GL-18 connections for integration with circulators and process tubing; suitable as a like-for-like replacement for common benchtop bioreactor systems.
  • 7 L nominal volume with 200 mL graduation subdivisions and numbered marks every 1,000 mL.
  • Constructed from heavy-wall borosilicate glass for thermal shock resistance and visibility.
  • Jacketed design enables circulating fluid temperature control.
  • GL-18 glass thread connections for circulator; supplied with caps and plastic hose fittings.
  • Minimum working volume of 2 L.
  • Vessel inner diameter approximately 160 mm and overall height approximately 375 mm.
  • Intended for laboratory-scale fermentation and bioprocess applications.
This item is not returnable. View return policy
Catalog No. NC2157841 Supplier Chemglass Life Sciences Supplier No. CLS138401
The Chemglass Jacketed Glass Bioreactor Vessel is a 7-liter jacketed borosilicate glass vessel designed for laboratory bioprocessing and fermentation. It provides jacketed temperature control, clear graduated sightlines, and GL-18 connections for integration with circulators and process tubing; suitable as a like-for-like replacement for common benchtop bioreactor systems.
  • 7 L nominal volume with 200 mL graduation subdivisions and numbered marks every 1,000 mL.
  • Constructed from heavy-wall borosilicate glass for thermal shock resistance and visibility.
  • Jacketed design enables circulating fluid temperature control.
  • GL-18 glass thread connections for circulator; supplied with caps and plastic hose fittings.
  • Minimum working volume of 2 L.
  • Vessel inner diameter approximately 160 mm and overall height approximately 375 mm.
  • Intended for laboratory-scale fermentation and bioprocess applications.