Learn More
Chemglass Life Sciences Tripod Stand for 7L Jacketed Bio Reactor Vessels
SDP
Catalog No. NC2964486
Encompass
Tripod stand designed to support 7 L jacketed glass bioreactor vessels. The stand features an anodized aluminum top two-piece ring and electropolished stainless steel lower ring and legs for corrosion resistance and stability. Includes silicone O-ring and knurled nuts for head plate attachment.
  • Top two-piece ring made of anodized aluminum.
  • Lower ring and legs constructed from electropolished stainless steel.
  • Compatible with 7 L jacketed bioreactor vessels.
  • Includes silicone O-ring and six knurled nuts for head plate attachment.
  • Provides stable support for jacketed glass vessels during mixing and temperature-controlled processing.
Catalog No. NC2964486 Supplier Chemglass Life Sciences Supplier No. CLS1384STD7
Tripod stand designed to support 7 L jacketed glass bioreactor vessels. The stand features an anodized aluminum top two-piece ring and electropolished stainless steel lower ring and legs for corrosion resistance and stability. Includes silicone O-ring and knurled nuts for head plate attachment.
  • Top two-piece ring made of anodized aluminum.
  • Lower ring and legs constructed from electropolished stainless steel.
  • Compatible with 7 L jacketed bioreactor vessels.
  • Includes silicone O-ring and six knurled nuts for head plate attachment.
  • Provides stable support for jacketed glass vessels during mixing and temperature-controlled processing.