Learn More
Vetequip Inc 7L VENTED INDUCTION CHAMBER
SDP
Catalog No. NC1619199
Encompass_Preferred
7-Liter Vented Induction Chamber Fresh gas inlet-standard 15mm x 22mm Waste gas outlet-30mm OD
Catalog No. NC1619199 Supplier Vetequip Inc Supplier No. 941447
7-Liter Vented Induction Chamber Fresh gas inlet-standard 15mm x 22mm Waste gas outlet-30mm OD