Learn More
Polyscience 7L Space-Saving Refrigerated Circulator, Advanced Digital Controller (-40 to 200°C); 120V, 60Hz
SDP
Catalog No. NC1777048
Encompass_Preferred
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorAdvanced Digital Controller-40 to 200°C Working Temperature RangeTemperature Stability: ±0.01°Cooling Capacity at 20°C: 360W1100W HeaterMaximum Suction: 4.3 psi (0.30 bar)Flow Rate: 5.3 gpm (20.1 LPM)120V, 60HzFeatures:• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, • RS-232/RS-485, and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability
Catalog No. NC1777048 Supplier Polyscience Supplier No. AD07R40A11B
Add to Cart
Add to Cart
Request a Quote
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorAdvanced Digital Controller-40 to 200°C Working Temperature RangeTemperature Stability: ±0.01°Cooling Capacity at 20°C: 360W1100W HeaterMaximum Suction: 4.3 psi (0.30 bar)Flow Rate: 5.3 gpm (20.1 LPM)120V, 60HzFeatures:• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, • RS-232/RS-485, and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability