Learn More
Cayman Chemical 7-methoxy Apigeninidin (chloride), Purity: ≥98%, CAS: 161773-51-9, 10 mg
SDP
Catalog No. 502982855
Encompass_Preferred
A 3-deoxyanthocyanin; active against radial growth of G. sorghi and spore germination of P. oryzae (IC50 = 50 ppm for both)
Catalog No. 50-298-2855 Supplier Cayman Chemical Supplier No. 1975710 MG
A 3-deoxyanthocyanin; active against radial growth of G. sorghi and spore germination of P. oryzae (IC50 = 50 ppm for both)