Learn More
Cayman Chemical 7-Methoxy-4-(trifluoromethyl)coumarin, Purity: ≥95%, CAS: 575-04-2, 100 mg
SDP
Catalog No. 502950166
Encompass_Preferred
A fluorogenic substrate for CYPs; HFC is released upon enzymatic cleavage by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-295-0166 Supplier Cayman Chemical Supplier No. 34398100 MG
A fluorogenic substrate for CYPs; HFC is released upon enzymatic cleavage by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively