Learn More
Cayman Chemical 7Methoxy8 3morpholIn4yl 10mg
SDP
Catalog No. 502990328
Encompass_Preferred
A building block; has been used in the synthesis of PI3K inhibitors
Catalog No. 50-299-0328 Supplier Cayman Chemical Supplier No. 3094410 MG
A building block; has been used in the synthesis of PI3K inhibitors