Learn More
Alkali Scientific 7mL 10% Neutral Buffered Formalin (NBF) Specimen Containers, Pre Filled, Dual Click Leak Resistant Lids, 95 kPa Compliant, 100/Tray
SDP
Catalog No. 5000662995
Encompass_Preferred
These 7 mL 10% Neutral Buffered Formalin (NBF) Specimen Containers are pre filled and ready for immediate use in histology, pathology, and clinical laboratory environments. Designed for safety, reliability, and regulatory compliance, each container features a secure dual click lid that provides audible and tactile confirmation of proper closure to help ensure leak resistant transport and specimen integrity. Manufactured for dependable laboratory performance, these containers are fully 95 kPa compliant for safe specimen transport and are suitable for tissue collection, biopsy handling, and routine fixation procedures. The convenient 100 per tray packaging supports efficient workflow in high volume laboratories, hospitals, and research facilities, helping streamline specimen collection while reducing handling time and exposure risk.
Catalog No. 50-006-62995 Supplier Alkali Scientific Supplier No. NBF07
These 7 mL 10% Neutral Buffered Formalin (NBF) Specimen Containers are pre filled and ready for immediate use in histology, pathology, and clinical laboratory environments. Designed for safety, reliability, and regulatory compliance, each container features a secure dual click lid that provides audible and tactile confirmation of proper closure to help ensure leak resistant transport and specimen integrity. Manufactured for dependable laboratory performance, these containers are fully 95 kPa compliant for safe specimen transport and are suitable for tissue collection, biopsy handling, and routine fixation procedures. The convenient 100 per tray packaging supports efficient workflow in high volume laboratories, hospitals, and research facilities, helping streamline specimen collection while reducing handling time and exposure risk.