Learn More
S2 Media 7mL Antibiotic Medium #19 Slants, 16x105mm PP Self-Standing Tube, 50/box
Catalog No. 502995885
Encompass_Preferred
Antibiotic Medium #19 is recommended for determining antibiotic potency of Amphotericin B, Candicidin, and Nystatin via cylinder plate (plate) and/or turbidimetric (tube) methods, as specified in the applicable compendium. First described by Kirshbaum and Arret, this formulation provides essential nutrients and other factors required for growth of Saccharomyces species, including S. cerevisiae and S. kudriavzevii.• Product meets EP, USP-NF, and WHO reference standards and performance criteria.• Antibiotic Medium #19 should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5885 Supplier S2 Media Supplier No. 5490
Antibiotic Medium #19 is recommended for determining antibiotic potency of Amphotericin B, Candicidin, and Nystatin via cylinder plate (plate) and/or turbidimetric (tube) methods, as specified in the applicable compendium. First described by Kirshbaum and Arret, this formulation provides essential nutrients and other factors required for growth of Saccharomyces species, including S. cerevisiae and S. kudriavzevii.• Product meets EP, USP-NF, and WHO reference standards and performance criteria.• Antibiotic Medium #19 should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.