Learn More
Leica Biosystems 7ml CD34 Bond RTU Primary
Supplier:  Leica Biosystems PA0212
Encompass_Preferred
Antigen BackgroundThe CD34 antigen is a single chain transmembrane glycoprotein with a molecular weight of 11 kD. The CD34 protein is selectively expressed on human lymphoid and myeloid hemopoietic progenitor cells. The CD34 antigen is also expressed on vascular enothelium.Product Specific InformationEnzyme digestion of paraffin sections is recommended with clone QBEnd/1 in perference to heat induced epitope retrieval as it produces stronger staining and reduces background elastin stainingDisclaimerCD34 (Endothelial Cell Marker) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2133