Learn More
Leica Biosystems 7ml CD45RO Bond RTU Primary
Supplier:  Leica Biosystems PA0146
Encompass
Antigen BackgroundThe CD45RO molecule, a 18 kD isoform of CD45, is reported to be expressed on 48% of peripheral blood T lymphocytes, 37% of CD4 positive lymphocytes, 8% of thymocytes and on the majority of T cell malignancies. Monocytes and granulocytes show surface expression of the antigen whereas tissue macrophages exhibit cytoplasmic expression.DisclaimerCD45RO is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2148