Learn More
Leica Biosystems 7ML Glypican-3 (1G12 Mab) 'CM'
Supplier:  Leica Biosystems PA0800
Encompass
This antibody is intended for in vitro diagnostic (IVD) use. The Cell Marque Glypican-3 (1G12) antibody is intended for qualified laboratories to qualitatively identify by light microscopy the presence of associated antigens in sections of formalin-fixed, paraffin-embedded tissue sections using IHC test methods. Use of this antibody is indicated, subsequent to clinical differential diagnosis, as an aid in the identification of hepatocellular carcinoma within the context of an antibody panel, the patient’s clinical history, and other diagnostic tests evaluated by a qualified pathologist.
Catalog No. 50-255-2040