Learn More
Leica Biosystems 7ml HMB45 Bond RTU Primary
Supplier:  Leica Biosystems PA0027U
Encompass
Antigen BackgroundThe HMB45 antigen has also been identified in retinal pigment epithelium (RPE) but is reported to be reactive only with the transient prenatal and infantile RPE. No reaction is reported to be observed with intradermal nevi and normal adult melanocytes and non-melanocytic cells.Tumor cells of epithelial, lymphoid, glial and mesenchymal origin are reported to be negative. This clone is well described in the literature. It is indicated to label an intracytoplasmic antigen in the majority of melanomas and other tumors demonstrating melanoma/melanocytic differentiation.Product Specific InformationThe clone is also reported to react with junctional and blue nevus cells. (Bacchi CE et al., A Review. Applied Immunohistochemistry. 4:73-85 (1996)).DisclaimerMelanoma Marker (HMB45) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2180